Assay Details
| Gene Symbol: FSD1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003480570.4 | XP_003480618.1 | 453 | 73 | ||
| XM_005660313.3 | XP_005660370.1 | 404 | 73 | |||
| XM_005660314.3 | XP_005660371.1 | 404 | 73 | |||
More Information
Set Membership: |
|

FSD1L
Genomic Map