Assay Details
| Gene Symbol: STK40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095939.1 | XP_020951598.1 | 1216 | 58 | ||
| XM_021095940.1 | XP_020951599.1 | 1452 | 58 | |||
| XM_021095941.1 | XP_020951600.1 | 1073 | 58 | |||
More Information
Set Membership: |
|

STK40
Genomic Map