Assay Details
| Gene Symbol: MAFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124249.6 | XP_003124297.1 | 295 | 122 | ||
| XM_013987694.2 | XP_013843148.1 | 198 | 122 | |||
| XM_013987695.2 | XP_013843149.1 | 1201 | 122 | |||
| XM_021085966.1 | XP_020941625.1 | 106 | 122 | |||
More Information
Set Membership: |
|

MAFK
Genomic Map