Assay Details
Gene Symbol: KIAA0895L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021094032.1 | XP_020949691.1 | 2417 | 68 | ||
XM_021094033.1 | XP_020949692.1 | 1188 | 68 | |||
XM_021094034.1 | XP_020949693.1 | 1576 | 68 | |||
XM_021094035.1 | XP_020949694.1 | 1012 | 68 | |||
XM_021094036.1 | XP_020949695.1 | 429 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
