Assay Details
| Gene Symbol: BTN2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656220.3 | XP_005656277.2 | 917 | 93 | ||
| XM_013988947.2 | XP_013844401.2 | 763 | 93 | |||
| XM_021099805.1 | XP_020955464.1 | 748 | 93 | |||
More Information
Set Membership: |
|

BTN2A2
Genomic Map