Assay Details
| Gene Symbol: EFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079969.1 | XP_020935628.1 | 2253 | 61 | ||
| XM_021079970.1 | XP_020935629.1 | 545 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: EFNB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079969.1 | XP_020935628.1 | 2253 | 61 | ||
| XM_021079970.1 | XP_020935629.1 | 545 | 61 | |||
Set Membership: |
|