Assay Details
| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123535.6 | XP_003123583.3 | 686 | 92 | ||
| XM_005652922.3 | XP_005652979.1 | 732 | 92 | |||
| XM_021083472.1 | XP_020939131.1 | 672 | 92 | |||
More Information
Set Membership: |
|

CERS1
Genomic Map