Assay Details
| Gene Symbol: PTPDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124787.4 | XP_003124835.1 | 391 | 69 | ||
| XM_003124788.4 | XP_003124836.2 | 2864 | 69 | |||
| XM_005662265.3 | XP_005662322.1 | 557 | 69 | |||
| XM_013995795.2 | XP_013851249.1 | 2864 | 69 | |||
| XM_021086999.1 | XP_020942658.1 | 2864 | 69 | |||
| XM_021087000.1 | XP_020942659.1 | 2864 | 69 | |||
More Information
Set Membership: |
|

PTPDC1
Genomic Map