Assay Details
| Gene Symbol: ACER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354020.3 | XP_003354068.1 | 490 | 67 | ||
| XM_021083989.1 | XP_020939648.1 | 649 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ACER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003354020.3 | XP_003354068.1 | 490 | 67 | ||
| XM_021083989.1 | XP_020939648.1 | 649 | 67 | |||
Set Membership: |
|