Assay Details
| Gene Symbol: MAFF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924465.4 | XP_001924500.1 | 320 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001924465.4 transcripts.
Set Membership: |
|

MAFF
Genomic Map