Assay Details
| Gene Symbol: ZNF496 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123573.6 | XP_003123621.3 | 332 | 90 | ||
| XM_013987506.2 | XP_013842960.1 | 334 | 90 | |||
| XM_013987507.2 | XP_013842961.1 | 333 | 90 | |||
| XM_021083404.1 | XP_020939063.1 | 335 | 90 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003123573.6, XM_013987506.2, XM_013987507.2, XM_021083404.1 transcripts.
Set Membership: |
|

ZNF496
Genomic Map