Assay Details
Gene Symbol: LOC100513450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005673619.2 | XP_005673676.1 | 294 | 80 | ||
XM_005673620.2 | XP_005673677.1 | 298 | 80 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100513450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005673619.2 | XP_005673676.1 | 294 | 80 | ||
XM_005673620.2 | XP_005673677.1 | 298 | 80 |