Assay Details
Gene Symbol: LOC100513450 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003135085.4 | XP_003135133.1 | 106 | 90 | ||
XM_005673620.2 | XP_005673677.1 | 106 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003135085.4, XM_005673620.2 transcripts.
Set Membership: |
![]() ![]() |
