Assay Details
| Gene Symbol: LOC100517427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483321.4 | XP_003483369.1 | 899 | 102 | ||
| XM_021070386.1 | XP_020926045.1 | 989 | 102 | |||
More Information
Set Membership: |
|

LOC100517427
Genomic Map