Assay Details
| Gene Symbol: ADAMTS5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070864.1 | XP_020926523.1 | 2973 | 108 | ||
| XM_021070865.1 | XP_020926524.1 | 3310 | 108 | |||
More Information
Set Membership: |
|

ADAMTS5
Genomic Map