Assay Details
| Gene Symbol: DOC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067535.1 | XP_020923194.1 | 950 | 66 | ||
| XM_021067536.1 | XP_020923195.1 | 606 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DOC2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021067535.1 | XP_020923194.1 | 950 | 66 | ||
| XM_021067536.1 | XP_020923195.1 | 606 | 66 | |||
Set Membership: |
|