Assay Details
| Gene Symbol: CROCC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021075704.1 | XP_020931363.1 | 6470 | 92 | ||
| XM_021075705.1 | XP_020931364.1 | 6461 | 92 | |||
| XM_021075706.1 | XP_020931365.1 | 6470 | 92 | |||
| XM_021075707.1 | XP_020931366.1 | 6470 | 92 | |||
More Information
Set Membership: |
|

CROCC2
Genomic Map