Assay Details
| Gene Symbol: CADM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929177.7 | XP_001929212.4 | 776 | 72 | ||
| XM_003481427.4 | XP_003481475.2 | 693 | 72 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CADM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929177.7 | XP_001929212.4 | 776 | 72 | ||
| XM_003481427.4 | XP_003481475.2 | 693 | 72 | |||
Set Membership: |
|