Assay Details
| Gene Symbol: CCSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021100737.1 | XP_020956396.1 | 2694 | 68 | ||
| XM_021100739.1 | XP_020956398.1 | 7898 | 68 | |||
| XM_021100740.1 | XP_020956399.1 | 4662 | 68 | |||
| XM_021100741.1 | XP_020956400.1 | 2616 | 68 | |||
More Information
Set Membership: |
|

CCSER1
Genomic Map