Assay Details
| Gene Symbol: LY6K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088773.1 | XP_020944432.1 | 235 | 67 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LY6K | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021088773.1 | XP_020944432.1 | 235 | 67 | ||