Assay Details
| Gene Symbol: STPG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021095544.1 | XP_020951203.1 | 1097 | 71 | ||
| XM_021095545.1 | XP_020951204.1 | 1355 | 71 | |||
| XM_021095546.1 | XP_020951205.1 | 1034 | 71 | |||
| XM_021095547.1 | XP_020951206.1 | 966 | 71 | |||
| XM_021095548.1 | XP_020951207.1 | 1094 | 71 | |||
| XM_021095549.1 | XP_020951208.1 | 1034 | 71 | |||
| XM_021095550.1 | XP_020951209.1 | 924 | 71 | |||
| XM_021095551.1 | XP_020951210.1 | 863 | 71 | |||
More Information
Set Membership: |
|

STPG1
Genomic Map