Assay Details
| Gene Symbol: TAFA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091913.1 | XP_020947572.1 | 1786 | 101 | ||
| XM_021091914.1 | XP_020947573.1 | 1899 | 101 | |||
| XM_021091915.1 | XP_020947574.1 | 391 | 101 | |||
More Information
Set Membership: |
|

TAFA2
Genomic Map