Assay Details
| Gene Symbol: PFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992193.2 | XP_013847647.1 | 674 | 101 | ||
| GenBank mRNA | JX457348.1 | - | 1 | 121 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PFN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013992193.2 | XP_013847647.1 | 674 | 101 | ||
| GenBank mRNA | JX457348.1 | - | 1 | 121 | 101 | |
Set Membership: |
|