Assay Details
| Gene Symbol: SLC13A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013981236.2 | XP_013836690.2 | 482 | 68 | ||
| XM_021067712.1 | XP_020923371.1 | 551 | 68 | |||
| GenBank mRNA | KF318030.1 | - | 769 | 68 | ||
| KF728381.1 | - | 3 - 4 | 769 | 68 | ||
More Information
Set Membership: |
|

SLC13A5
Genomic Map