Assay Details
| Gene Symbol: NMU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129032.4 | XP_003129080.1 | 205 | 65 | ||
| GenBank mRNA | KJ622334.1 | - | 1 - 2 | 112 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: NMU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129032.4 | XP_003129080.1 | 205 | 65 | ||
| GenBank mRNA | KJ622334.1 | - | 1 - 2 | 112 | 65 | |
Set Membership: |
|