Assay Details
| Gene Symbol: SEPTIN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086365.1 | XP_020942024.1 | 2017 | 112 | ||
| XM_021086366.1 | XP_020942025.1 | 1108 | 112 | |||
| GenBank mRNA | KU358532.1 | - | 5 - 6 | 725 | 112 | |
More Information
Set Membership: |
|

SEPTIN14
Genomic Map