Assay Details
| Gene Symbol: SEPTIN14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021086366.1 | XP_020942025.1 | 1509 | 107 | ||
| GenBank mRNA | KU358532.1 | - | 8 - 9 | 1126 | 107 | |
More Information
Set Membership: |
|

SEPTIN14
Genomic Map