Assay Details
| Gene Symbol: DAZL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358321.3 | XP_003358369.1 | 1474 | 57 | ||
| GenBank mRNA | KU705632.1 | - | 4 - 5 | 357 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: DAZL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003358321.3 | XP_003358369.1 | 1474 | 57 | ||
| GenBank mRNA | KU705632.1 | - | 4 - 5 | 357 | 57 | |
Set Membership: |
|