Assay Details
| Gene Symbol: IMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021074943.1 | XP_020930602.1 | 803 | 60 | ||
| XM_021074944.1 | XP_020930603.1 | 694 | 60 | |||
| XM_021074945.1 | XP_020930604.1 | 709 | 60 | |||
| GenBank mRNA | AK233589.1 | - | 5 - 6 | 492 | 60 | |
| AK234208.1 | - | 5 - 6 | 578 | 60 | ||
| AK393101.1 | - | 5 - 6 | 548 | 60 | ||
More Information
Set Membership: |
|

IMP4
Genomic Map