Assay Details
| Gene Symbol: EPM2AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233630.1 | - | 1 | 1521 | 87 | |
| AK234907.1 | - | 1 | 1362 | 87 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EPM2AIP1| Gene Symbol: EPM2AIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233630.1 | - | 1 | 1521 | 87 | |
| AK234907.1 | - | 1 | 1362 | 87 | ||
Set Membership: |
|