Assay Details
| Gene Symbol: VIRMA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021089096.1 | XP_020944755.1 | 4320 | 102 | ||
| XM_021089097.1 | XP_020944756.1 | 4202 | 102 | |||
| XM_021089098.1 | XP_020944757.1 | 4290 | 102 | |||
| XM_021089099.1 | XP_020944758.1 | 5537 | 102 | |||
| XM_021089100.1 | XP_020944759.1 | 5596 | 102 | |||
| GenBank mRNA | AK233640.1 | - | 8 - 9 | 1664 | 102 | |
| AK237935.1 | - | 2 - 3 | 273 | 102 | ||
More Information
Set Membership: |
|

VIRMA
Genomic Map