Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021079233.1 | XP_020934892.1 | 547 | 66 | ||
| GenBank mRNA | AK233699.1 | - | 1 - 2 | 132 | 66 | |
More Information
Set Membership: |
|

MINDY4
Genomic Map