Assay Details
| Gene Symbol: RPL8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005655297.3 | XP_005655354.1 | 616 | 52 | ||
| GenBank mRNA | AK233757.1 | - | 4 - 5 | 532 | 52 | |
| AK239846.1 | - | 4 - 5 | 728 | 52 | ||
| AK391946.1 | - | 4 - 5 | 533 | 52 | ||
| AK394824.1 | - | 4 - 5 | 561 | 52 | ||
More Information
Set Membership: |
|

RPL8
Genomic Map