Assay Details
| Gene Symbol: IL10RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656637.3 | XP_005656694.1 | 777 | 77 | ||
| XM_021062907.1 | XP_020918566.1 | 611 | 77 | |||
| GenBank mRNA | AK233769.1 | - | 5 - 6 | 774 | 77 | |
More Information
Set Membership: |
|

IL10RA
Genomic Map