Assay Details
| Gene Symbol: ZDHHC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK233786.1 | - | 1 | 920 | 129 | |
| AK240395.1 | - | 1 | 911 | 129 | ||
| AK390310.1 | - | 1 | 1512 | 129 | ||
More Information
Set Membership: |
|

ZDHHC1
Genomic Map