Assay Details
| Gene Symbol: RSRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483417.4 | XP_003483465.1 | 547 | 66 | ||
| XM_005670608.3 | XP_005670665.1 | 563 | 66 | |||
| XM_005670609.3 | XP_005670666.1 | 641 | 66 | |||
| XM_013982805.2 | XP_013838259.1 | 577 | 66 | |||
| XM_013982806.2 | XP_013838260.1 | 563 | 66 | |||
| XM_021074086.1 | XP_020929745.1 | 1065 | 66 | |||
| GenBank mRNA | AK233795.1 | - | 4 - 5 | 496 | 66 | |
More Information
Set Membership: |
|

RSRC2
Genomic Map