Assay Details
| Gene Symbol: KIFC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001927845.5 | XP_001927880.3 | 2164 | 61 | ||
| XM_021098594.1 | XP_020954253.1 | 2126 | 61 | |||
| XM_021098595.1 | XP_020954254.1 | 2124 | 61 | |||
| XM_021098596.1 | XP_020954255.1 | 2124 | 61 | |||
| XM_021098598.1 | XP_020954257.1 | 2482 | 61 | |||
| XM_021098599.1 | XP_020954258.1 | 3939 | 61 | |||
| GenBank mRNA | AK233842.1 | - | 10 - 11 | 2116 | 61 | |
More Information
Set Membership: |
|

KIFC1
Genomic Map