Assay Details
| Gene Symbol: PACSIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021091420.1 | XP_020947079.1 | 1656 | 62 | ||
| XM_021091421.1 | XP_020947080.1 | 1582 | 62 | |||
| XM_021091422.1 | XP_020947081.1 | 1530 | 62 | |||
| XM_021091423.1 | XP_020947082.1 | 1540 | 62 | |||
| XM_021091424.1 | XP_020947083.1 | 1437 | 62 | |||
| GenBank mRNA | AK233921.1 | - | 6 - 7 | 761 | 62 | |
More Information
Set Membership: |
|

PACSIN2
Genomic Map