Assay Details
| Gene Symbol: VKORC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124473.5 | XP_003124521.1 | 393 | 118 | ||
| XM_021086403.1 | XP_020942062.1 | 237 | 118 | |||
| GenBank mRNA | AK233937.1 | - | 1 - 2 | 238 | 118 | |
| AK345850.1 | - | 1 - 2 | 200 | 118 | ||
| AK347315.1 | - | 1 - 2 | 226 | 118 | ||
| AK391833.1 | - | 1 - 2 | 247 | 118 | ||
More Information
Set Membership: |
|

VKORC1
Genomic Map