Assay Details
| Gene Symbol: MAP2K4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132006.5 | XP_003132054.2 | 4803 | 67 | ||
| XM_021067890.1 | XP_020923549.1 | 572 | 67 | |||
| GenBank mRNA | AK233982.1 | - | 2 - 3 | 247 | 67 | |
More Information
Set Membership: |
|

MAP2K4
Genomic Map