Assay Details
| Gene Symbol: HSD17B14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094705.1 | XP_020950364.1 | 336 | 73 | ||
| GenBank mRNA | AK233987.1 | - | 2 - 3 | 322 | 73 | |
More Information
Set Membership: |
|

HSD17B14
Genomic Map