Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003126863.6 | XP_003126911.3 | 460 | 100 | ||
| XM_013998172.2 | XP_013853626.1 | 593 | 100 | |||
| GenBank mRNA | AK233991.1 | - | 2 - 3 | 450 | 100 | |
More Information
Set Membership: |
|

TMEM170A
Genomic Map