Assay Details
| Gene Symbol: SNUPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929236.5 | XP_001929271.1 | 911 | 72 | ||
| XM_003128480.4 | XP_003128528.1 | 833 | 72 | |||
| GenBank mRNA | AK234136.1 | - | 8 - 9 | 838 | 72 | |
| AK350475.1 | - | 7 - 8 | 755 | 72 | ||
More Information
Set Membership: |
|

SNUPN
Genomic Map