Assay Details
| Gene Symbol: LITAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124593.4 | XP_003124641.1 | 1736 | 102 | ||
| XM_021086604.1 | XP_020942263.1 | 1687 | 102 | |||
| XM_021086605.1 | XP_020942264.1 | 1873 | 102 | |||
| XM_021086606.1 | XP_020942265.1 | 1689 | 102 | |||
| XM_021086607.1 | XP_020942266.1 | 1688 | 102 | |||
| XM_021086608.1 | XP_020942267.1 | 1687 | 102 | |||
| XM_021086609.1 | XP_020942268.1 | 1687 | 102 | |||
| GenBank mRNA | AK234150.1 | - | 1 | 912 | 102 | |
| AK394083.1 | - | 4 | 1702 | 102 | ||
| AK395663.1 | - | 4 | 1705 | 102 | ||
| AK398580.1 | - | 1 | 1029 | 102 | ||
More Information
Set Membership: |
|

LITAF
Genomic Map