Assay Details
| Gene Symbol: FAAP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021097589.1 | XP_020953248.1 | 1845 | 55 | ||
| GenBank mRNA | AK234151.1 | - | 3 - 4 | 550 | 55 | |
More Information
Set Membership: |
|

FAAP20
Genomic Map