Assay Details
| Gene Symbol: TMEM222 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482003.4 | XP_003482051.1 | 367 | 58 | ||
| GenBank mRNA | AK234174.1 | - | 3 - 4 | 211 | 58 | |
| AK391169.1 | - | 3 - 4 | 211 | 58 | ||
More Information
Set Membership: |
|

TMEM222
Genomic Map