Assay Details
| Gene Symbol: NAGK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125043.6 | XP_003125091.3 | 456 | 58 | ||
| XM_005662479.3 | XP_005662536.1 | 513 | 58 | |||
| XM_005662480.3 | XP_005662537.1 | 517 | 58 | |||
| XM_021087390.1 | XP_020943049.1 | 501 | 58 | |||
| GenBank mRNA | AK234234.1 | - | 4 - 5 | 389 | 58 | |
| AK236476.1 | - | 5 - 6 | 481 | 58 | ||
| AK389299.1 | - | 4 - 5 | 397 | 58 | ||
| AY610103.1 | - | 4 - 5 | 377 | 58 | ||
More Information
Set Membership: |
|

NAGK
Genomic Map