Assay Details
| Gene Symbol: SVBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021096916.1 | XP_020952575.1 | 576 | 105 | ||
| GenBank mRNA | AK234245.1 | - | 1 - 2 | 516 | 105 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021096916.1 transcripts.
Set Membership: |
|

SVBP
Genomic Map