Assay Details
| Gene Symbol: LOC100157391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926555.6 | XP_001926590.3 | 643 | 142 | ||
| XM_005671220.3 | XP_005671277.1 | 568 | 142 | |||
| GenBank mRNA | AK234276.1 | - | 5 - 6 | 525 | 142 | |
| AK235825.1 | - | 5 - 6 | 566 | 142 | ||
| AK392422.1 | - | 5 - 6 | 530 | 142 | ||
| AK400657.1 | - | 5 - 6 | 571 | 142 | ||
| AY610146.1 | - | 5 - 6 | 648 | 142 | ||
More Information
Set Membership: |
|

LOC100157391
Genomic Map