Assay Details
| Gene Symbol: LOC100157391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001926555.6 | XP_001926590.3 | 759 | 95 | ||
| GenBank mRNA | AK234276.1 | - | 6 - 7 | 641 | 95 | |
| AK392422.1 | - | 6 - 7 | 646 | 95 | ||
| AK400657.1 | - | 6 - 7 | 687 | 95 | ||
| AY610146.1 | - | 6 - 7 | 764 | 95 | ||
More Information
Set Membership: |
|

LOC100157391
Genomic Map